siRNA Details
Gene Symbol: Sap18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009119.3 | NP_033145.2 | 2 | 260 | ||
GenBank mRNA | AK005189.1 | - | 2 | 178 | ||
AK006647.1 | - | 2 | 199 | |||
AK085992.1 | - | 2 | 199 | |||
AK133619.1 | - | 2 | 194 | |||
AK144904.1 | - | 2 | 233 | |||
AK146928.1 | - | 2 | 233 | |||
AK209620.1 | - | 1 | 17 | |||
BC006625.1 | - | 2 | 201 | 3968600 | ||
BC099480.1 | - | 2 | 194 | 30934194 | ||
CF730347.1 | - | 2 | 260 | |||
Z97062.1 | - | 2 | 195 |
Gene Symbol: Gm10094 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142441.1 | NP_001135913.1 | 1 | 231 | ||
GenBank mRNA | C88706.1 | - | 1 | 236 |

More Information
Other Species Targeted

Gene Ontology Categories:
