siRNA Details
Gene Symbol: Ccl21b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011335.2 | NP_035465.2 | 157 | |||
GenBank mRNA | W67046.1 | - | 2 | 157 |
Gene Symbol: Ccl21a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011124.4 | NP_035254.1 | 2 | 194 | ||
GenBank mRNA | AF001980.1 | - | 2 | 121 | ||
AF006637.1 | - | 2 | 217 | |||
AK144258.1 | - | 2 | 196 | |||
AK155924.1 | - | 2 | 194 | |||
BC025974.1 | - | 2 | 193 | 4208773 | ||
BC028747.1 | - | 2 | 181 | 1196234 | ||
BC038120.1 | - | 2 | 163 | 3372243 |
Gene Symbol: Ccl21c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023052.2 | NP_075539.1 | 2 | 193 | ||
GenBank mRNA | BC087957.1 | - | 2 | 193 | 6750979 | |
U88322.1 | - | 2 | 166 |
Gene Symbol: Gm10591 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193668.1 | NP_001180597.1 | 193 |
Gene Symbol: Gm13304 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193666.1 | NP_001180595.1 | 193 |
Gene Symbol: Gm1987 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193667.1 | NP_001180596.1 | 2 | 196 |
Gene Symbol: Gm21541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270360.1 | NP_001257289.1 | 2 | 193 | ||
GenBank mRNA | BX522471.1 | - | 2 | 214 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
