siRNA Details
Gene Symbol: Ccl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009139.3 | NP_033165.1 | 2, 3 | 268 | ||
GenBank mRNA | AF128188.1 | - | 2, 3 | 164 | ||
AF128189.1 | - | 2, 3 | 164 | |||
AF128190.1 | - | 2, 3 | 164 | |||
AF128191.1 | - | 2, 3 | 164 | |||
AF128192.1 | - | 2, 3 | 164 | |||
AK002697.1 | - | 2, 3 | 171 | |||
AK008547.1 | - | 2, 3 | 172 | |||
AK150336.1 | - | 2, 3 | 171 | |||
AK154553.1 | - | 2, 3 | 172 | |||
BY172106.1 | - | 2, 3 | 268 | |||
M58004.1 | - | 2, 3 | 171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
