siRNA Details
Gene Symbol: Nhp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011482.4 | NP_035612.2 | 3 | 1231 | ||
GenBank mRNA | AK004489.1 | - | 3 | 1179 | ||
AK134943.1 | - | 1169 | ||||
AK145391.1 | - | 3 | 1233 | |||
AK145608.1 | - | 3 | 1213 | |||
AK168930.1 | - | 3 | 1184 | |||
AK169103.1 | - | 3 | 1216 | |||
BC026755.1 | - | 3 | 1177 | 3154066 | ||
BC054450.1 | - | 3 | 1177 | 6491298 | ||
BC083315.1 | - | 3 | 1153 | 5714819 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
