siRNA Details
Gene Symbol: Stat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011486.4 | NP_035616.1 | 9 | 1090 | ||
NM_213659.2 | NP_998824.1 | 9 | 1090 | |||
NM_213660.2 | NP_998825.1 | 9 | 1090 | |||
GenBank mRNA | AK145486.1 | - | 9 | 946 | ||
AK153005.1 | - | 9 | 1002 | |||
AK153472.1 | - | 9 | 1002 | |||
AK197148.1 | - | 2 | 76 | |||
AY299489.1 | - | 8 | 817 | |||
AY299490.1 | - | 8 | 817 | |||
BC003806.1 | - | 9 | 978 | 3593588 | ||
BC019168.1 | - | 9 | 994 | 4923137 | ||
L29278.1 | - | 9 | 1069 | |||
U06922.1 | - | 9 | 879 | |||
U08378.1 | - | 9 | 957 | |||
U30709.1 | - | 9 | 983 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
