siRNA Details
Gene Symbol: Gm3258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011509.2 | NP_035639.1 | 1 | 463 |
Gene Symbol: Supt4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009296.1 | NP_033322.1 | 3 | 228 | ||
GenBank mRNA | AK002990.1 | - | 3 | 330 | ||
AK146235.1 | - | 4 | 385 | |||
AK167941.1 | - | 3 | 242 | |||
AK168229.1 | - | 4 | 385 | |||
BC024391.1 | - | 3 | 223 | 5011064 | ||
BC057949.1 | - | 200 | ||||
BC061174.1 | - | 3 | 229 | 6592996 | ||
BC087923.1 | - | 3 | 204 | 30308013 | ||
BC141092.1 | - | 3 | 266 | 9055704 | ||
U43154.1 | - | 3 | 193 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
