siRNA Details
Gene Symbol: Zfp719 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172482.1 | NP_766070.1 | 4 | 991 | ||
GenBank mRNA | AK033195.1 | - | 1 | 370 | ||
AK044684.1 | - | 4 | 968 | |||
AK083143.1 | - | 4 | 932 | |||
BC081548.1 | - | 4 | 954 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
