siRNA Details
Gene Symbol: BC048679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193274.1 | NP_001180203.1 | 6 | 651 | ||
NM_183143.3 | NP_898966.1 | 6 | 533 | |||
GenBank mRNA | AY226992.1 | - | 6 | 484 | ||
BB073717.1 | - | 2 | 194 | |||
BC048679.1 | - | 6 | 602 |
