siRNA Details
Gene Symbol: Vmn2r112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104575.1 | NP_001098045.1 | 3 | 745 | ||
GenBank mRNA | AB540943.1 | - | 3 | 745 |
Gene Symbol: Vmn2r111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104573.1 | NP_001098043.1 | 3 | 745 | ||
GenBank mRNA | AB540944.1 | - | 3 | 745 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
