siRNA Details
Gene Symbol: A730008H23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172505.4 | NP_766093.1 | 5 | 1892 | ||
GenBank mRNA | AK042589.1 | - | 5 | 1892 | ||
AK138428.1 | - | 5 | 1855 | |||
BC131919.1 | - | 5 | 1860 | 40130196 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
