siRNA Details
Gene Symbol: Ago3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153402.2 | NP_700451.2 | 18 | 2622 | ||
GenBank mRNA | AB081473.1 | - | 18 | 2474 | ||
AK035042.1 | - | 18 | 2586 | |||
AK083652.1 | - | 18 | 2590 | |||
AK154436.1 | - | 18 | 2623 | |||
AK171118.1 | - | 18 | 2623 | |||
BC137964.1 | - | 18 | 2469 | 8860983 | ||
BC145061.1 | - | 18 | 2446 | |||
BC145063.1 | - | 17 | 2332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
