siRNA Details
Gene Symbol: Trp53i13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024920.1 | NP_001020091.1 | 7 | 1302 | ||
GenBank mRNA | AK003215.1 | - | 2 | 572 | ||
BC062271.1 | - | 1 | 610 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
