siRNA Details
Gene Symbol: Synrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194341.2 | NP_919322.1 | 11, 12 | 1402 | ||
GenBank mRNA | BC056370.1 | - | 11, 12 | 1362 | 6831893 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
