siRNA Details
Gene Symbol: Synrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194341.2 | NP_919322.1 | 3 | 229 | ||
GenBank mRNA | BC056370.1 | - | 3 | 189 | 6831893 | |
BY135827.1 | - | 3 | 233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
