siRNA Details
Gene Symbol: Prr15l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146026.1 | NP_666138.1 | 3 | 462 | ||
GenBank mRNA | AK077279.1 | - | 2 | 435 | ||
AK087566.1 | - | 314 | ||||
AK090284.1 | - | 2 | 464 | |||
BC022765.1 | - | 3 | 462 | 4925830 | ||
BC024510.1 | - | 2 | 346 | 4983784 | ||
BC034192.1 | - | 2 | 392 | 4984606 | ||
BC034213.1 | - | 2 | 441 | 4989688 | ||
BC036171.1 | - | 2 | 409 | 4988054 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
