siRNA Details
Gene Symbol: Tmc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195088.1 | NP_001182017.1 | 8 | 1261 | ||
NM_001195089.1 | NP_001182018.1 | 8 | 1160 | |||
NM_001195090.1 | NP_001182019.1 | 8 | 1157 | |||
NM_181856.2 | NP_862904.1 | 8 | 1157 | |||
GenBank mRNA | AK143862.1 | - | 8 | 1265 | ||
AK156764.1 | - | 8 | 1118 | |||
AK171772.1 | - | 8 | 1143 | |||
AK220211.1 | - | 5 | 629 | |||
AY236501.1 | - | 8 | 1069 | |||
AY263160.1 | - | 7 | 1045 | |||
BC138254.1 | - | 7 | 1033 | 8861273 | ||
BC145210.1 | - | 7 | 1036 | 9053761 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
