siRNA Details
Gene Symbol: Ska3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198605.3 | NP_941007.1 | 4 | 488 | ||
GenBank mRNA | AK089223.1 | - | 4 | 489 | ||
AK158142.1 | - | 4 | 510 | |||
AK166171.1 | - | 4 | 479 | |||
BC117500.1 | - | 4 | 480 | 40105921 | ||
BC117501.1 | - | 4 | 480 | 40105920 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
