siRNA Details
Gene Symbol: Clec3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011606.2 | NP_035736.2 | 3 | 894 | ||
GenBank mRNA | BC035043.1 | - | 3 | 846 | 1313548 | |
U08595.1 | - | 3 | 879 | |||
X79199.1 | - | 3 | 867 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
