siRNA Details
Gene Symbol: Rpl13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009438.5 | NP_033464.2 | 7 | 536 | ||
GenBank mRNA | AK007609.1 | - | 7 | 526 | ||
AK077512.1 | - | 7 | 528 | |||
AK146318.1 | - | 7 | 528 | |||
AK156035.1 | - | 7 | 528 | |||
AK159882.1 | - | 7 | 526 | |||
AK168784.1 | - | 7 | 528 | |||
AK168903.1 | - | 7 | 528 | |||
AK170030.1 | - | 7 | 439 | |||
AK213140.1 | - | 2 | 157 | |||
BC034450.1 | - | 1 | 69 | |||
BC042392.1 | - | 1 | 69 | |||
BC065799.1 | - | 4 | 1099 | |||
BC082289.1 | - | 7 | 512 | 6486177 | ||
BC086896.1 | - | 7 | 528 | 6776320 | ||
CT010169.1 | - | 6 | 505 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
