siRNA Details
Gene Symbol: Tuba1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009448.4 | NP_033474.1 | 4 | 1906 | ||
GenBank mRNA | AK013029.1 | - | 2 | 1062 | ||
AK145567.1 | - | 4 | 1506 | |||
AK167654.1 | - | 4 | 1476 | |||
AK167759.1 | - | 4 | 1439 | |||
AK191962.1 | - | 1 | 284 | |||
BC004745.1 | - | 4 | 1451 | 3583832 | ||
BC022182.1 | - | 4 | 1516 | 3661220 | ||
BC026753.1 | - | 4 | 1431 | 5124686 | ||
BC094022.1 | - | 4 | 1906 | 4162287 | ||
M13441.1 | - | 4 | 1418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
