siRNA Details
Gene Symbol: Ung | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040691.1 | NP_001035781.1 | 7 | 1429 | ||
NM_011677.2 | NP_035807.2 | 6 | 1401 | |||
GenBank mRNA | AK146178.1 | - | 7 | 1435 | ||
AK168709.1 | - | 6 | 1351 | |||
AK168785.1 | - | 6 | 1351 | |||
BC004037.1 | - | 5 | 1222 | |||
BC011039.1 | - | 7 | 1476 | 4009947 | ||
BC052853.1 | - | 6 | 1338 | 30058519 | ||
U55041.1 | - | 6 | 1353 | |||
X99018.1 | - | 6 | 1352 | |||
Y08975.1 | - | 7 | 1424 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
