siRNA Details
Gene Symbol: Lin7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011699.3 | NP_035829.1 | 4 | 421 | ||
GenBank mRNA | AF087695.1 | - | 4 | 421 | ||
AF173083.1 | - | 4 | 428 | |||
AK015399.1 | - | 4 | 440 | |||
AK078923.1 | - | 4 | 441 | |||
AK083299.1 | - | 4 | 445 | |||
AK128989.1 | - | 4 | 441 | |||
AK133531.1 | - | 4 | 430 | |||
AK135810.1 | - | 4 | 447 | |||
AK161076.1 | - | 4 | 402 | |||
AK162185.1 | - | 2 | 266 | |||
BC046966.1 | - | 4 | 421 | 6493071 | ||
BC052471.1 | - | 4 | 431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
