siRNA Details
Gene Symbol: Eny2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175009.3 | NP_778174.1 | 4 | 303 | ||
GenBank mRNA | AF173297.1 | - | 4 | 323 | ||
AK007877.1 | - | 4 | 254 | |||
AK016562.1 | - | 4 | 1688 | |||
AK032957.1 | - | 4 | 303 | |||
AK160628.1 | - | 4 | 269 | |||
AK162511.1 | - | 4 | 298 | |||
BC048361.1 | - | 4 | 309 | 4012930 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001130580, XM_006241615, FQ223243.
Gene Ontology Categories:
