siRNA Details
Gene Symbol: Senp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144851.5 | NP_659100.1 | 15, 16 | 2006 | ||
GenBank mRNA | AK028565.1 | - | 15, 16 | 1748 | ||
AK028762.1 | - | 7, 8 | 2980 | |||
AK035723.1 | - | 16, 17 | 2189 | |||
AK054521.1 | - | 16, 17 | 2138 | |||
AK088597.1 | - | 15, 16 | 1750 | |||
BC023129.1 | - | 14, 15 | 1720 | 3595534 |

More Information
Other Species Targeted

Gene Ontology Categories:
