siRNA Details
Gene Symbol: Rbbp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081223.2 | NP_001074692.1 | 18, 19 | 3027 | ||
NM_001252495.1 | NP_001239424.1 | 18, 19 | 2932 | |||
GenBank mRNA | AK037049.1 | - | 18, 19 | 2950 | ||
AK041384.1 | - | 17, 18 | 2777 | |||
AK077614.1 | - | 17, 18 | 2979 | |||
AK163908.1 | - | 2867 | ||||
BC019665.1 | - | 15, 16 | 2372 | |||
BC050849.1 | - | 18, 19 | 2932 | 6508067 | ||
CX567593.1 | - | 4, 5 | 464 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
