siRNA Details
Gene Symbol: Fhod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289654.1 | NP_001276583.1 | 19 | 4226 | ||
NM_001289655.1 | NP_001276584.1 | 20 | 4583 | |||
NM_175276.4 | NP_780485.2 | 21 | 4679 | |||
GenBank mRNA | AB078608.1 | - | 21 | 3857 | ||
AB078609.1 | - | 19 | 3404 | |||
AB100088.1 | - | 20 | 3761 | |||
AK148245.1 | - | 19 | 3368 | |||
AK220413.1 | - | 17 | 2843 | |||
BC056371.1 | - | 11 | 2311 | |||
BC060607.1 | - | 6 | 1635 | |||
BC138046.1 | - | 19 | 3326 | 8861065 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
