siRNA Details
Gene Symbol: Piezo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039485.4 | NP_001034574.4 | 47 | 8192 | ||
GenBank mRNA | AK031235.1 | - | 7 | 1130 | ||
AK031422.1 | - | 6 | 851 | |||
BC026614.1 | - | 4 | 580 | |||
BC075677.1 | - | 8 | 1283 | |||
BC147606.1 | - | 6 | 745 | 9056596 | ||
BC172109.1 | - | 6 | 745 | |||
HQ215521.1 | - | 47 | 7272 |
