siRNA Details
Gene Symbol: Farp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145519.2 | NP_663494.2 | 25 | 2920 | ||
GenBank mRNA | AK050860.1 | - | 25 | 2885 | ||
AK171713.1 | - | 25 | 2946 | |||
AK173028.1 | - | 2 | 176 | |||
AW912106.1 | - | 2 | 191 | |||
BC009153.1 | - | 25 | 2909 | 2655209 |
