siRNA Details
Gene Symbol: Gm128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024841.3 | NP_001020012.1 | 1 | 1123 | ||
GenBank mRNA | AB690575.1 | - | 1 | 995 | ||
AK144488.1 | - | 1 | 1124 | |||
BC092540.1 | - | 1 | 1220 | 6742207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
