siRNA Details
Gene Symbol: Tnfrsf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178931.2 | NP_849262.1 | 8 | 821 | ||
GenBank mRNA | AF515707.1 | - | 8 | 793 | ||
AY264405.1 | - | 8 | 821 | |||
BC022125.1 | - | 6 | 555 | |||
BC104054.1 | - | 8 | 821 | 40040309 | ||
BC104055.1 | - | 8 | 824 | 40040311 |
