siRNA Details
Gene Symbol: Pilra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153510.3 | NP_705730.1 | 2 | 620 | ||
GenBank mRNA | AJ400844.1 | - | 518 | |||
AK041413.1 | - | 1 | 911 | |||
AK041670.1 | - | 2 | 532 | |||
AK170000.1 | - | 2 | 532 | |||
BC115565.2 | - | 505 | 40102015 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
