siRNA Details
Gene Symbol: Zbtb45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024699.1 | NP_001019870.1 | 3 | 1470 | ||
GenBank mRNA | AK154877.1 | - | 3 | 1548 | ||
AK155880.1 | - | 3 | 1532 | |||
AK166382.1 | - | 1539 | ||||
AK183668.1 | - | 2 | 93 | |||
AK190203.1 | - | 2 | 93 | |||
BC094359.1 | - | 3 | 1470 | 6828341 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
