siRNA Details
Gene Symbol: Wdr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199060.1 | NP_001185989.1 | 3, 4 | 666 | ||
NM_001199061.1 | NP_001185990.1 | 3, 4 | 398 | |||
NM_021312.5 | NP_067287.1 | 2, 3 | 488 | |||
GenBank mRNA | AB041608.1 | - | 3, 4 | 667 | ||
AF239765.1 | - | 2, 3 | 144 | |||
AK007500.1 | - | 2, 3 | 265 | |||
AK012022.1 | - | 2, 3 | 242 | |||
AK016611.1 | - | 3, 4 | 342 | |||
AK031436.1 | - | 2, 3 | 251 | |||
AK149085.1 | - | 2, 3 | 170 | |||
AK161299.1 | - | 3, 4 | 411 | |||
AK213584.1 | - | 1, 2 | 37 | |||
AU080254.1 | - | 3, 4 | 666 | |||
AY059431.1 | - | 2, 3 | 179 | |||
BC004748.1 | - | 2, 3 | 322 | 3583955 | ||
BC052386.1 | - | 2, 3 | 459 | 6418498 | ||
BC096651.1 | - | 2, 3 | 233 | 1514703 | ||
CX564842.1 | - | 2, 3 | 489 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
