siRNA Details
Gene Symbol: BC030336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164580.1 | NP_001158052.1 | 2 | 294 | ||
GenBank mRNA | AK052350.1 | - | 2 | 145 | ||
BC030336.1 | - | 2 | 178 | 5401060 |

More Information
Other Species Targeted

Gene Ontology Categories:
