siRNA Details
Gene Symbol: Ddx60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293783.1 | NP_001280712.1 | 37 | 5290 | ||
GenBank mRNA | AK136403.1 | - | 4 | 2370 | ||
BC013672.1 | - | 10 | 1278 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ddx60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293783.1 | NP_001280712.1 | 37 | 5290 | ||
GenBank mRNA | AK136403.1 | - | 4 | 2370 | ||
BC013672.1 | - | 10 | 1278 |