siRNA Details
Gene Symbol: Tmem205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253867.1 | NP_001240796.1 | 3 | 419 | ||
NM_001253868.1 | NP_001240797.1 | 3 | 492 | |||
NM_001253869.1 | NP_001240798.1 | 3 | 351 | |||
NM_001253870.1 | NP_001240799.1 | 3 | 479 | |||
NM_178577.5 | NP_848692.2 | 3 | 487 | |||
GenBank mRNA | AB030201.1 | - | 3 | 322 | ||
AI573565.1 | - | 3 | 303 | |||
AK161954.1 | - | 1 | 1537 | |||
BC010787.1 | - | 3 | 414 | 4211629 | ||
CA326029.1 | - | 3 | 351 | |||
CN840781.1 | - | 3 | 407 |
