siRNA Details
Gene Symbol: D630033O11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243261.1 | NP_001230190.1 | 3 | 370 | ||
GenBank mRNA | AK052711.1 | - | 2 | 227 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
