siRNA Details
Gene Symbol: Fscn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172802.4 | NP_766390.2 | 1 | 731 | ||
GenBank mRNA | AK044556.1 | - | 1 | 730 | ||
AK050002.1 | - | 1 | 732 | |||
AK078674.1 | - | 1 | 759 | |||
BC109356.1 | - | 1 | 692 | 40057445 | ||
BC109357.1 | - | 1 | 692 | 40057447 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
