siRNA Details
Gene Symbol: Gpr68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177673.1 | NP_001171144.1 | 3 | 377 | ||
NM_001177674.1 | NP_001171145.1 | 3 | 559 | |||
NM_175493.4 | NP_780702.1 | 3 | 680 | |||
GenBank mRNA | AK036659.1 | - | 2 | 389 | ||
AK087410.1 | - | 3 | 361 | |||
AK154649.1 | - | 2 | 352 | |||
AK155886.1 | - | 2 | 369 | |||
BC118034.1 | - | 2 | 358 | 40106885 | ||
BC118044.1 | - | 2 | 358 | 40106886 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
