siRNA Details
Gene Symbol: Hax1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282032.1 | NP_001268961.1 | 4 | 504 | ||
NM_011826.4 | NP_035956.1 | 5 | 756 | |||
GenBank mRNA | AF023482.1 | - | 5 | 728 | ||
AF023483.1 | - | 4 | 891 | |||
AF465243.1 | - | 4 | 368 | |||
AK002256.1 | - | 5 | 707 | |||
AK010633.1 | - | 5 | 740 | |||
AK088746.1 | - | 5 | 738 | |||
AK135138.1 | - | 4 | 466 | |||
BC006688.1 | - | 5 | 685 | 3582652 | ||
BC098225.1 | - | 5 | 710 | 1494110 | ||
BC147481.1 | - | 5 | 687 | 9056226 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
