siRNA Details
Gene Symbol: Gucy1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204340.1 | NP_001191269.1 | 4 | 473 | ||
NM_172810.3 | NP_766398.1 | 4 | 473 | |||
GenBank mRNA | AK047129.1 | - | 4 | 474 | ||
AY236513.1 | - | 2 | 147 | |||
BC137868.1 | - | 4 | 470 | 8860887 | ||
BC144988.1 | - | 4 | 470 | 9053536 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
