siRNA Details
Gene Symbol: Oas1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145211.2 | NP_660212.2 | 4 | 879 | ||
GenBank mRNA | AK149613.1 | - | 4 | 840 | ||
BC013715.1 | - | 4 | 861 | 3257027 | ||
BC057878.1 | - | 4 | 838 | 5348858 | ||
M33863.1 | - | 865 | ||||
X04958.1 | - | 4 | 865 |
Gene Symbol: Oas1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF480417.1 | - | 4 | 866 | ||
BC018470.1 | - | 850 | 3493001 | |||
BC043339.1 | - | 4 | 942 | 5004267 | ||
X58077.1 | - | 793 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
