siRNA Details
Gene Symbol: Dazap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011873.2 | NP_036003.2 | 4 | 503 | ||
GenBank mRNA | AF085348.1 | - | 4 | 468 | ||
AK002595.1 | - | 4 | 540 | |||
AK050159.1 | - | 4 | 517 | |||
AK088916.1 | - | 4 | 525 | |||
AK132139.1 | - | 4 | 517 | |||
AK137246.1 | - | 4 | 504 | |||
AK145398.1 | - | 4 | 537 | |||
AK146970.1 | - | 4 | 528 | |||
AK151730.1 | - | 1 | 3308 | |||
AK152511.1 | - | 1 | 3307 | |||
BC014808.1 | - | 4 | 503 | 4506124 | ||
BC029351.1 | - | 3 | 434 | |||
BC083347.1 | - | 4 | 495 | 6829350 |

More Information
Other Species Targeted

Gene Ontology Categories:
