siRNA Details
Gene Symbol: Arid1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085355.1 | NP_001078824.1 | 2 | 1440 | ||
GenBank mRNA | AK160334.1 | - | 1 | 166 | ||
BF462904.1 | - | 2 | 93 |

More Information
Other Species Targeted

Gene Ontology Categories:
