siRNA Details
Gene Symbol: Neurl1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081656.2 | NP_001075125.1 | 3 | 673 | ||
GenBank mRNA | DQ839448.1 | - | 3 | 673 | ||
GQ414760.1 | - | 3 | 673 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
