siRNA Details
Gene Symbol: Dmxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081371.2 | NP_001074840.2 | 24 | 6156 | ||
GenBank mRNA | AY590892.1 | - | 24 | 6156 | ||
BC056490.1 | - | 1 | 320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
