siRNA Details
Gene Symbol: Ccl19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011888.2 | NP_036018.1 | 4 | 631 | ||
GenBank mRNA | AF059208.2 | - | 4 | 631 | ||
AK144337.1 | - | 4 | 640 | |||
AK156269.1 | - | 1941 | ||||
BC025130.1 | - | 612 | ||||
BC051472.1 | - | 621 | 1349213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
