siRNA Details
Gene Symbol: Ccdc85b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243307.1 | NP_001230236.1 | 2 | 704 | ||
NM_198616.4 | NP_941018.1 | 1 | 859 | |||
GenBank mRNA | AK140354.1 | - | 1 | 1193 | ||
BC016659.1 | - | 2 | 644 | |||
BC058411.1 | - | 1 | 845 | 5710206 | ||
BC094592.1 | - | 2 | 645 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
