siRNA Details
Gene Symbol: Rgsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243223.1 | NP_001230152.1 | 16 | 2773 | ||
GenBank mRNA | AK029630.1 | - | 7 | 880 | ||
AK161463.1 | - | 9 | 1193 |

More Information
Other Species Targeted

Gene Ontology Categories:
