siRNA Details
Gene Symbol: Tnfsf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183391.3 | NP_899247.3 | 3 | 415 | ||
GenBank mRNA | AJ577579.1 | - | 3 | 417 | ||
AJ577580.1 | - | 417 | ||||
AY234223.1 | - | 3 | 373 | |||
AY267900.1 | - | 3 | 359 | |||
AY320040.1 | - | 3 | 365 | |||
AY359852.1 | - | 3 | 359 | |||
BC137814.1 | - | 3 | 415 | 8860833 | ||
BC137815.1 | - | 3 | 415 | 8860834 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
